Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC103689989 SIRNA Assays ›
          Gene Symbol:
          LOC103689989 LOC103690150 Prl3d1 Prl3d2 Prl3d4
          Gene Name:
          prolactin-3D4
          prolactin-3D1
          Prolactin family 3, subfamily d, member 1
          Prolactin family 3, subfamily d, member 2
          prolactin family 3, subfamily d, member 4
          Gene Aliases:
          -
          -
          Csh1, Pl-I, Pl-Im, Pl1, Rhco1, rPl-1
          Pl-1
          Csh1l1, Csh1v, Pl-Iv
          Chromosome Location:
          Chr.17: 42920887 - 42927745 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 39721993 - 39728391 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 39777255 - 39782877 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 42849469 - 42855494 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 39271883 - 39278716 on Build Rnor_6.0
          UniGene ID:
          -
          Rn.112569
          Rn.112569
          Rn.198338
          Rn.1259
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID s132386

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC103689989
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008771673.1 XP_008769895.1 150
          XM_008771674.1 XP_008769896.1 188
          Gene Symbol: LOC103690150
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008771672.1 XP_008769894.1 185
          GenBank mRNA FQ228618.1 - 208
          Gene Symbol: Prl3d1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006253914.2 2 208
          GenBank mRNA FQ220053.1 - 212
          FQ220072.1 - 208
          FQ220274.1 - 210
          FQ220568.1 - 210
          FQ220587.1 - 208
          FQ221231.1 - 210
          FQ223300.1 - 211
          FQ228787.1 - 208
          FQ229328.1 - 208
          FQ229537.1 - 207
          FQ229656.1 - 202
          FQ230119.1 - 208
          M55269.1 - 201
          Gene Symbol: Prl3d2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008771671.1 XP_008769893.1 2 211
          GenBank mRNA BC161851.1 - 2 185 9088452
          D21103.1 - 2 147
          DQ329282.1 - 2 204
          DQ329283.1 - 2 204
          FQ145043.1 - 2 210
          FQ221365.1 - 2 208
          FQ223255.1 - 2 211
          FQ229928.1 - 2 210
          FQ230035.1 - 2 208
          Gene Symbol: Prl3d4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033233.1 NP_150236.1 2 168
          XM_008771595.1 XP_008769817.1 2 150
          GenBank mRNA BC097302.1 - 2 194 7444366
          U32679.1 - 2 168

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          activation of Janus kinase activity
          hormone activity
          prolactin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0