Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Phgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031620.1 | NP_113808.1 | 6 | 608 | ||
XM_006233010.1 | XP_006233072.1 | 5 | 631 | |||
GenBank mRNA | BC086327.1 | - | 5 | 636 | 7115765 | |
X97772.1 | - | 5 | 567 |