Search Thermo Fisher Scientific
Gene Symbol: Igbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031624.3 | NP_113812.1 | 4 | 748 | ||
GenBank mRNA | AF000577.2 | - | 4 | 740 | ||
BC068202.1 | - | 4 | 752 | 6921265 | ||
FQ212697.1 | - | 4 | 757 | |||
FQ222671.1 | - | 4 | 750 |