Search
Search

Gpr85| Gene Symbol: Gpr85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022254.3 | NP_071590.1 | 2 | 751 | ||
| XM_006236124.2 | XP_006236186.1 | 4 | 1690 | |||
| XM_006236126.2 | XP_006236188.1 | 3 | 1566 | |||
| XM_008762724.1 | XP_008760946.1 | 3 | 1481 | |||
| GenBank mRNA | AB040803.1 | - | 1 | 489 | ||
| AF203907.1 | - | 1 | 513 | |||
| BC087727.1 | - | 2 | 751 | 7320496 | ||
Set Membership: |
|