Search
Search

Basp1
LOC100910172| Gene Symbol: Basp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022300.1 | NP_071636.1 | 2 | 981 | ||
| GenBank mRNA | D14441.1 | - | 2 | 981 | ||
| Gene Symbol: LOC100910172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750615.3 | XP_003750663.2 | 1 | 1217 | ||