Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SRPK2 SIRNA Assays ›
          Gene Symbol:
          SRPK2
          Gene Name:
          SRSF protein kinase 2
          Gene Aliases:
          SFRSK2
          Chromosome Location:
          Chr.7: 105110708 - 105399366 on Build GRCh38
          UniGene ID:
          Hs.285197
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13454

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SRPK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278273.1 NP_001265202.1 13 1750
          NM_182691.2 NP_872633.1 11 1803
          NM_182692.2 NP_872634.1 12 1726
          XM_005250549.3 XP_005250606.2 13 1985
          XM_005250550.3 XP_005250607.1 12 1730
          XM_005250551.4 XP_005250608.1 13 1778
          XM_006716098.2 XP_006716161.1 13 1745
          XM_011516536.2 XP_011514838.1 11 1741
          XM_011516538.2 XP_011514840.1 12 1614
          XM_017012560.1 XP_016868049.1 13 1800
          XM_017012561.1 XP_016868050.1 12 1650
          XM_017012562.1 XP_016868051.1 13 1984
          XM_017012563.1 XP_016868052.1 13 1985
          XM_017012564.1 XP_016868053.1 11 1789
          XM_017012565.1 XP_016868054.1 13 1749
          XM_017012566.1 XP_016868055.1 13 1786
          GenBank mRNA AK093420.1 - 3 1525
          AY354201.1 - 12 1791
          BC035214.1 - 11 1722 4778251
          BC068547.1 - 13 1750 5265599

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001106575, XM_006235903, XM_008762636, XM_008762637, XM_008762635, and others.
          This siRNA also targets the following transcripts in Mouse: NM_009274, AK035962, BC062941, AK041426, AB006036, and others.

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal complex assembly
          angiogenesis
          protein phosphorylation
          positive regulation of cell proliferation
          RNA splicing
          positive regulation of gene expression
          cell differentiation
          nuclear speck organization
          intracellular signal transduction
          positive regulation of neuron apoptotic process
          positive regulation of viral genome replication
          negative regulation of viral genome replication
          innate immune response
          positive regulation of cell cycle
          regulation of mRNA splicing, via spliceosome
          magnesium ion binding
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          poly(A) RNA binding
          14-3-3 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0