Search
Search

Apaf1| Gene Symbol: Apaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023979.1 | NP_076469.1 | 9 | 1873 | ||
| XM_008765263.1 | XP_008763485.1 | 6 | 1665 | |||
| GenBank mRNA | AF218388.1 | - | 9 | 1328 | ||
| AF320222.1 | - | 9 | 1873 | |||