Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024159.1 | NP_077073.1 | 5 | 483 | ||
GenBank mRNA | AF045658.1 | - | 2 | 130 | ||
BC097314.1 | - | 5 | 518 | 7444786 | ||
FQ224185.1 | - | 793 | ||||
U95177.1 | - | 5 | 483 | |||
U95178.1 | - | 5 | 483 |