Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST5 SIRNA Assays ›
Gene Symbol:
ST5
Gene Name:
suppression of tumorigenicity 5
Gene Aliases:
DENND2B, HTS1, p126
Chromosome Location:
Chr.11: 8693352 - 8910951 on Build GRCh38
UniGene ID:
Hs.117715
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13514
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005418.3
NP_005409.3
20
3352
NM_139157.2
NP_631896.1
16
1820
NM_213618.1
NP_998783.1
17
3152
XM_005253077.1
XP_005253134.1
17
3067
XM_005253083.2
XP_005253140.1
16
1962
XM_011520309.1
XP_011518611.1
18
3127
XM_011520310.1
XP_011518612.1
18
3539
XM_011520311.2
XP_011518613.1
18
3521
XM_011520312.1
XP_011518614.1
22
3486
XM_011520313.1
XP_011518615.1
21
3372
XM_011520314.1
XP_011518616.1
20
3264
XM_011520315.1
XP_011518617.1
20
3360
XM_011520316.1
XP_011518618.1
19
3290
XM_011520317.1
XP_011518619.1
18
3231
XM_011520318.1
XP_011518620.1
21
3426
XM_011520319.2
XP_011518621.1
18
3079
XM_011520323.2
XP_011518625.2
17
1950
XM_011520324.2
XP_011518626.2
16
1890
XM_011520325.1
XP_011518627.1
17
2022
XM_011520326.1
XP_011518628.1
21
2226
XM_011520327.1
XP_011518629.1
17
1915
XM_011520328.1
XP_011518630.1
17
1947
XM_011520329.1
XP_011518631.1
20
2166
XM_017018182.1
XP_016873671.1
19
3245
XM_017018183.1
XP_016873672.1
19
3222
XM_017018184.1
XP_016873673.1
21
3381
XM_017018185.1
XP_016873674.1
17
3177
XM_017018186.1
XP_016873675.1
16
3117
XM_017018187.1
XP_016873676.1
17
1886
GenBank mRNA
AK127763.1
-
14
2207
AK293245.1
-
14
1585
AK307617.1
-
6
707
AK312305.1
-
17
3118
AK312758.1
-
16
2162
BC036655.1
-
20
3357
4837231
BX647431.1
-
17
3152
S45936.1
-
13
1490
U15131.1
-
17
3080
U15779.1
-
13
1449
U15780.1
-
16
1820
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of GTPase activity
Rab guanyl-nucleotide exchange factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0