Search
Search

Creb1
LOC100363500| Gene Symbol: Creb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031017.1 | NP_112279.1 | 9 | 940 | ||
| NM_134443.1 | NP_604392.1 | 7 | 859 | |||
| XM_006245065.2 | XP_006245127.1 | 9 | 946 | |||
| XM_006245066.2 | XP_006245128.1 | 8 | 1111 | |||
| XM_006245067.2 | XP_006245129.1 | 8 | 851 | |||
| XM_006245068.2 | XP_006245130.1 | 7 | 798 | |||
| XM_006245069.2 | XP_006245131.1 | 7 | 1448 | |||
| XM_006245070.2 | XP_006245132.1 | 4 | 318 | |||
| XM_008767138.1 | XP_008765360.1 | 6 | 3302 | |||
| XM_008767139.1 | XP_008765361.1 | 3 | 1172 | |||
| GenBank mRNA | BC097311.1 | - | 10 | 1204 | ||
| X14788.1 | - | 9 | 940 | |||
| X60002.1 | - | 7 | 859 | |||
| Gene Symbol: LOC100363500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772050.1 | XP_008770272.1 | 1, 2 | 894 | ||