Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TGFBR2 SIRNA Assays ›
Gene Symbol:
Tgfbr2
Gene Name:
transforming growth factor, beta receptor 2
Gene Aliases:
TGF-beta 2, Tgfbr2T
Chromosome Location:
Chr.8: 124310288 - 124399345 on Build Rnor_6.0
UniGene ID:
Rn.9954
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s135790
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tgfbr2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031132.3
NP_112394.3
1, 2
345
GenBank mRNA
GU067683.1
-
1, 2
94
L09653.1
-
1, 2
152
S67770.1
-
1, 2
345
Back To Top
More Information
Panther Classification:
Molecular Function -
serine/threonine protein kinase receptor
Gene Ontology Categories:
Function(s)
Process(es)
patterning of blood vessels
vasculogenesis
response to hypoxia
in utero embryonic development
positive regulation of mesenchymal cell proliferation
lens development in camera-type eye
positive regulation of tolerance induction to self antigen
positive regulation of B cell tolerance induction
positive regulation of T cell tolerance induction
growth plate cartilage development
growth plate cartilage chondrocyte growth
protein phosphorylation
receptor-mediated endocytosis
apoptotic process
transmembrane receptor protein serine/threonine kinase signaling pathway
transforming growth factor beta receptor signaling pathway
common-partner SMAD protein phosphorylation
smoothened signaling pathway
gastrulation
brain development
heart development
embryo implantation
aging
response to nutrient
negative regulation of cell proliferation
response to mechanical stimulus
response to glucose
organ morphogenesis
response to organic substance
regulation of gene expression
positive regulation of epithelial cell migration
response to organic cyclic compound
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
lung development
organ regeneration
activation of protein kinase activity
embryonic hemopoiesis
wound healing
regulation of cell proliferation
response to drug
myeloid dendritic cell differentiation
positive regulation of skeletal muscle tissue regeneration
response to estrogen
positive regulation of angiogenesis
response to steroid hormone
digestive tract development
positive regulation of smooth muscle cell proliferation
embryonic cranial skeleton morphogenesis
positive regulation of NK T cell differentiation
cartilage development
palate development
negative regulation of cardiac muscle cell proliferation
pathway-restricted SMAD protein phosphorylation
lung morphogenesis
bronchus development
bronchus morphogenesis
trachea morphogenesis
trachea formation
mammary gland morphogenesis
lung lobe morphogenesis
response to cholesterol
lens fiber cell apoptotic process
positive regulation of reactive oxygen species metabolic process
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
transforming growth factor beta-activated receptor activity
transforming growth factor beta receptor activity, type II
protein binding
ATP binding
glycosaminoglycan binding
mitogen-activated protein kinase kinase kinase binding
type I transforming growth factor beta receptor binding
type III transforming growth factor beta receptor binding
protein homodimerization activity
SMAD binding
metal ion binding
protein heterodimerization activity
transforming growth factor beta binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0