Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GNB2L1 SIRNA Assays ›
          Gene Symbol:
          Gnb2l1
          Gene Name:
          guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1
          Gene Aliases:
          RACK1
          Chromosome Location:
          Chr.10: 34149717 - 34155198 on Build Rnor_6.0
          UniGene ID:
          Rn.55487
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s135893

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gnb2l1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_130734.2 NP_570090.2 3 523
          GenBank mRNA BC063809.1 - 3 498 6919982
          FQ212638.1 - 3 494
          FQ213067.1 - 3 494
          FQ213571.1 - 3 495
          FQ215527.1 - 3 494
          FQ215812.1 - 3 494
          FQ216167.1 - 3 494
          FQ216212.1 - 3 493
          FQ218475.1 - 3 524
          FQ218650.1 - 3 498
          FQ219434.1 - 3 494
          FQ219492.1 - 3 495
          FQ219693.1 - 3 494
          FQ219729.1 - 3 494
          FQ219878.1 - 3 492
          FQ219900.1 - 3 494
          FQ220124.1 - 3 494
          FQ220151.1 - 3 494
          FQ220277.1 - 3 494
          FQ220314.1 - 3 494
          FQ220391.1 - 3 494
          FQ220419.1 - 3 494
          FQ220428.1 - 3 498
          FQ220430.1 - 3 493
          FQ220494.1 - 3 494
          FQ220585.1 - 3 494
          FQ220598.1 - 3 498
          FQ220697.1 - 3 498
          FQ220819.1 - 3 495
          FQ220976.1 - 3 501
          FQ221075.1 - 3 494
          FQ221140.1 - 3 501
          FQ221286.1 - 3 498
          FQ222502.1 - 3 494
          FQ223855.1 - 3 493
          FQ224952.1 - 3 494
          FQ225047.1 - 3 498
          FQ225312.1 - 3 493
          FQ225511.1 - 3 494
          FQ225843.1 - 3 524
          FQ226802.1 - 3 494
          FQ228007.1 - 3 498
          FQ229157.1 - 3 493
          FQ229247.1 - 3 494
          FQ229458.1 - 3 491
          FQ229569.1 - 3 495
          FQ229616.1 - 3 494
          FQ229633.1 - 3 494
          FQ229689.1 - 3 494
          FQ229758.1 - 3 494
          FQ229908.1 - 3 495
          FQ229911.1 - 3 494
          FQ229916.1 - 3 492
          FQ229923.1 - 3 493
          FQ230003.1 - 3 492
          FQ230018.1 - 3 502
          FQ230117.1 - 3 494
          FQ230328.1 - 3 495
          FQ233024.1 - 3 494
          FQ234000.1 - 3 500
          FQ234673.1 - 3 496
          FQ235255.1 - 3 494
          U03390.1 - 3 486

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          positive regulation of protein phosphorylation
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          protein kinase C-activating G-protein coupled receptor signaling pathway
          gastrulation
          negative regulation of gene expression
          negative regulation of translation
          negative regulation of Wnt signaling pathway
          negative regulation of cell growth
          positive regulation of cell migration
          positive regulation of cAMP catabolic process
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of protein homooligomerization
          regulation of protein localization
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          rhythmic process
          negative regulation of phagocytosis
          regulation of cell division
          positive regulation of cyclic-nucleotide phosphodiesterase activity
          regulation of cell cycle
          negative regulation of protein kinase B signaling
          positive regulation of mitochondrial depolarization
          negative regulation of protein tyrosine kinase activity
          regulation of establishment of protein localization to plasma membrane
          negative regulation of hydrogen peroxide-induced neuron death
          regulation of establishment of cell polarity
          positive regulation of gastrulation
          positive regulation of intrinsic apoptotic signaling pathway
          protein kinase C binding
          ion channel inhibitor activity
          cysteine-type endopeptidase activator activity involved in apoptotic process
          enzyme binding
          protein phosphatase binding
          protein tyrosine kinase inhibitor activity
          receptor tyrosine kinase binding
          SH2 domain binding
          protein homodimerization activity
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0