Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fcna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031348.2 | NP_112638.2 | 10 | 1016 | ||
XM_008761614.1 | XP_008759836.1 | 10 | 1049 | |||
GenBank mRNA | AB026057.2 | - | 10 | 927 | ||
BC088128.1 | - | 10 | 1016 | 7376326 |