Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Taldo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031811.3 | NP_113999.2 | 3, 4 | 352 | ||
GenBank mRNA | AF069306.1 | - | 3, 4 | 338 | ||
BC059126.1 | - | 3, 4 | 356 | 6889440 | ||
FQ213939.1 | - | 3, 4 | 364 | |||
FQ219168.1 | - | 3, 4 | 378 |