Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HDAC2 SIRNA Assays ›
Gene Symbol:
Hdac2
Gene Name:
histone deacetylase 2
Gene Aliases:
-
Chromosome Location:
Chr.20: 43084870 - 43108198 on Build Rnor_6.0
UniGene ID:
Rn.1797
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s136732
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hdac2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053447.1
NP_445899.1
2
312
GenBank mRNA
BC161939.1
-
2
287
DY318702.1
-
2
312
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
cardiac muscle hypertrophy
maintenance of chromatin silencing
transcription, DNA-templated
protein deacetylation
positive regulation of cell proliferation
epidermal cell differentiation
positive regulation of epithelial to mesenchymal transition
positive regulation of receptor biosynthetic process
negative regulation of neuron projection development
dendrite development
histone deacetylation
hippocampus development
neuron differentiation
response to caffeine
response to lipopolysaccharide
positive regulation of interleukin-1 production
positive regulation of tumor necrosis factor production
circadian regulation of gene expression
cellular response to heat
response to nicotine
response to cocaine
odontogenesis of dentin-containing tooth
response to drug
positive regulation of tyrosine phosphorylation of Stat3 protein
embryonic digit morphogenesis
ATP-dependent chromatin remodeling
negative regulation of apoptotic process
negative regulation of DNA binding
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of proteolysis
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
behavioral response to ethanol
positive regulation of oligodendrocyte differentiation
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of protein kinase B signaling
cardiac muscle cell development
response to hyperoxia
negative regulation of cardiac muscle cell proliferation
regulation of sarcomere organization
hair follicle placode formation
negative regulation of dendritic spine development
eyelid development in camera-type eye
fungiform papilla formation
histone H3-K9 modification
cellular response to hydrogen peroxide
histone H3 deacetylation
histone H4 deacetylation
cellular response to retinoic acid
cellular response to transforming growth factor beta stimulus
negative regulation of canonical Wnt signaling pathway
regulation of protein deacetylation
cellular response to dopamine
negative regulation of peptidyl-lysine acetylation
negative regulation of intrinsic apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
core promoter binding
RNA polymerase II repressing transcription factor binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
histone deacetylase activity
protein binding
transcription factor binding
deacetylase activity
enzyme binding
heat shock protein binding
chromatin DNA binding
nucleosomal DNA binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
Krueppel-associated box domain binding
sequence-specific DNA binding
poly(A) RNA binding
NAD-dependent histone deacetylase activity (H3-K9 specific)
NAD-dependent histone deacetylase activity (H4-K16 specific)
NAD-dependent histone deacetylase activity (H3-K18 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0