Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Jak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053466.1 | NP_445918.1 | 11 | 1739 | ||
XM_006238452.2 | XP_006238514.1 | 11 | 1912 | |||
XM_006238453.2 | XP_006238515.1 | 11 | 5535 | |||
GenBank mRNA | AJ000556.1 | - | 7 | 1217 |