Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC1 SIRNA Assays ›
Gene Symbol:
TACC1
Gene Name:
transforming acidic coiled-coil containing protein 1
Gene Aliases:
Ga55
Chromosome Location:
Chr.8: 38728186 - 38853028 on Build GRCh38
UniGene ID:
Hs.279245
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13717
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
392.00
Your Price
Online offer:
380.65
380.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122824.1
NP_001116296.1
7, 8
1120
NM_001146216.2
NP_001139688.1
9, 10
2296
NM_006283.2
NP_006274.2
9, 10
2350
XM_005273625.4
XP_005273682.1
11, 12
2500
XM_005273628.1
XP_005273685.1
10, 11
2007
XM_005273629.1
XP_005273686.1
11, 12
2020
XM_005273630.1
XP_005273687.1
10, 11
2028
XM_011544631.1
XP_011542933.1
10, 11
2386
XM_011544632.2
XP_011542934.1
12, 13
2561
XM_011544633.1
XP_011542935.1
10, 11
2299
XM_011544634.1
XP_011542936.1
9, 10
2263
XM_011544635.1
XP_011542937.1
10, 11
2090
XM_011544636.2
XP_011542938.1
7, 8
746
XM_011544637.1
XP_011542939.1
7, 8
1033
XM_011544638.2
XP_011542940.1
7, 8
636
XM_017013766.1
XP_016869255.1
9, 10
1971
XM_017013767.1
XP_016869256.1
10, 11
1984
XM_017013768.1
XP_016869257.1
9, 10
1884
XM_017013769.1
XP_016869258.1
10, 11
1897
XM_017013770.1
XP_016869259.1
9, 10
1920
XM_017013771.1
XP_016869260.1
9, 10
2192
XM_017013772.1
XP_016869261.1
8, 9
1884
XM_017013773.1
XP_016869262.1
8, 9
2151
XM_017013774.1
XP_016869263.1
8, 9
1836
XM_017013775.1
XP_016869264.1
9, 10
1905
XM_017013776.1
XP_016869265.1
10, 11
1897
XM_017013777.1
XP_016869266.1
8, 9
2061
XM_017013778.1
XP_016869267.1
8, 9
1760
GenBank mRNA
AF049910.1
-
9, 10
2291
AK294931.1
-
9, 10
1864
AK295071.1
-
9, 10
1898
AK304774.1
-
7, 8
670
AK314620.1
-
9, 10
2350
AY039239.1
-
8, 9
660
AY139007.1
-
1, 2
995
AY177411.1
-
7, 8
1061
BC041391.1
-
11, 12
2460
5276767
BX647696.1
-
9, 10
2295
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell cycle
cell proliferation
cerebral cortex development
cell division
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0