Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NRG1 SIRNA Assays ›
          Gene Symbol:
          Nrg1
          Gene Name:
          neuregulin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 62969988 - 64057434 on Build Rnor_6.0
          UniGene ID:
          Rn.37438
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s137273

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nrg1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271118.1 NP_001258047.1 6, 7 1739
          NM_001271119.1 NP_001258048.1 5, 6 1724
          NM_001271120.1 NP_001258049.1 5, 6 1724
          NM_001271121.1 NP_001258050.1 7, 8 1168
          NM_001271122.1 NP_001258051.1 8, 9 1192
          NM_001271124.1 NP_001258053.1 9, 10 1279
          NM_001271125.1 NP_001258054.1 10, 11 1348
          NM_001271126.1 NP_001258055.1 10, 11 1294
          NM_001271127.1 NP_001258056.1 10, 11 1348
          NM_001271128.1 NP_001258057.1 9, 10 1279
          NM_001271129.1 NP_001258058.1 9, 10 1279
          NM_001271130.1 NP_001258059.1 10, 11 1364
          NM_031588.2 NP_113776.2 9, 10 1270
          GenBank mRNA AF194438.1 - 6, 7 1753
          AF194439.1 - 5, 6 1472
          AF194440.1 - 5, 6 547
          AF194993.1 - 10, 11 1364
          AY973241.1 - 8, 9 684
          AY973243.1 - 7, 8 662
          AY973244.1 - 8, 9 650
          AY973245.1 - 10, 11 1015
          AY995221.1 - 10, 11 950
          AY995222.1 - 7, 8 824
          AY995223.1 - 8, 9 848
          M92430.1 - 9, 10 979
          U02316.1 - 9, 10 1279
          U02317.1 - 9, 10 1279
          U02318.1 - 9, 10 1270
          U02319.1 - 9, 10 1279
          U02321.1 - 9, 10 1270
          U02322.1 - 10, 11 1348
          U02323.1 - 9, 10 1279
          U02324.1 - 9, 10 979

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_013960, NM_001159995, NM_001160004, NM_013964, NM_013957, and others.

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          cell morphogenesis
          cardiac conduction system development
          ventricular trabecula myocardium morphogenesis
          transmembrane receptor protein tyrosine kinase signaling pathway
          activation of transmembrane receptor protein tyrosine kinase activity
          nervous system development
          synapse assembly
          peripheral nervous system development
          heart development
          muscle organ development
          locomotory behavior
          regulation of blood pressure
          cell proliferation
          glial cell differentiation
          negative regulation of cardiac muscle cell apoptotic process
          glucose transport
          cell migration
          glial cell fate commitment
          cell differentiation
          positive regulation of cell growth
          intracellular signal transduction
          ERBB signaling pathway
          wound healing
          negative regulation of protein catabolic process
          myelination
          regulation of protein heterodimerization activity
          cellular protein complex disassembly
          neurotransmitter receptor metabolic process
          regulation of cell differentiation
          positive regulation of cell adhesion
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          positive regulation of Ras protein signal transduction
          animal organ development
          neuron fate commitment
          negative regulation of secretion
          positive regulation of protein kinase B signaling
          ventricular cardiac muscle cell differentiation
          positive regulation of cardiac muscle cell proliferation
          cardiac muscle cell myoblast differentiation
          endocardial cell differentiation
          positive regulation of protein tyrosine kinase activity
          positive regulation of calcineurin-NFAT signaling cascade
          positive regulation of cardiac muscle cell differentiation
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          transcription cofactor activity
          receptor binding
          ErbB-2 class receptor binding
          growth factor activity
          protein tyrosine kinase activator activity
          ErbB-3 class receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qlr2r:80/100.66.77.194:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0