Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FCGR2A SIRNA Assays ›
          Gene Symbol:
          Fcgr2a LOC100911825 LOC103693683 LOC103694908 LOC498276
          Gene Name:
          Fc fragment of IgG, low affinity IIa, receptor
          low affinity immunoglobulin gamma Fc region receptor III-like
          low affinity immunoglobulin gamma Fc region receptor III
          low affinity immunoglobulin gamma Fc region receptor III-like
          Fc gamma receptor II beta
          Gene Aliases:
          Fcgr3, Fcgr3a
          -
          -
          -
          -
          Chromosome Location:
          Chr.13: 91146879 - 91163694 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 91219838 - 91237277 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 89420835 - 89440644 on Build Rnor_6.0
          Chromosome Location:
          Chr.Un|NW_007906965: 601 - 13578 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 91168972 - 91181954 on Build Rnor_6.0
          UniGene ID:
          Rn.154415
          -
          Rn.6050
          -
          Rn.6050
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID s138102

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fcgr2a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053843.2 NP_446295.2 6 905
          XM_006250303.2 XP_006250365.1 6 958
          XM_006250305.2 XP_006250367.1 6 1057
          GenBank mRNA BC060534.1 - 682 6886727
          L08446.1 - 6 905
          M32062.1 - 718
          M64368.1 - 698
          M64369.1 - 698
          M64370.1 - 698
          Gene Symbol: LOC100911825
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006250308.2 XP_006250370.1 5 726
          XM_008769774.1 XP_008767996.1 5 726
          XM_008769775.1 XP_008767997.1 6 951
          XM_008769776.1 XP_008767998.1 6 759
          XM_008769777.1 XP_008767999.1 4 705
          Gene Symbol: LOC103693683
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008769767.1 XP_008767989.1 6 747
          XM_008769768.1 XP_008767990.1 6 952
          XM_008769769.1 XP_008767991.1 6 1539
          XM_008769770.1 XP_008767992.1 6 1521
          XM_008769771.1 XP_008767993.1 5 728
          XM_008769772.1 XP_008767994.1 6 1132
          XM_008769773.1 XP_008767995.1 5 710
          Gene Symbol: LOC103694908
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008773904.1 XP_008772126.1 726
          XM_008773905.1 XP_008772127.1 727
          XM_008773906.1 XP_008772128.1 854
          Gene Symbol: LOC498276
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135992.1 NP_001129464.1 765
          XM_008769748.1 XP_008767970.1 726
          XM_008769749.1 XP_008767971.1 726
          XM_008769750.1 XP_008767972.1 850
          GenBank mRNA BC159405.1 - 765 7459664

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          nerve development
          Fc-gamma receptor signaling pathway
          IgG receptor activity
          IgG binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline