Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109911.1 | NP_001103381.1 | 12, 13 | 1136 | ||
NM_053876.2 | NP_446328.2 | 13, 14 | 1019 | |||
XM_006241116.2 | XP_006241178.1 | 12, 13 | 2326 | |||
XM_008765198.1 | XP_008763420.1 | 10, 11 | 866 | |||
GenBank mRNA | AY724543.1 | - | 12, 13 | 1123 | ||
BC088317.1 | - | 8, 9 | 647 | 7325202 | ||
U32577.2 | - | 13, 14 | 1019 |