Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIMS2 SIRNA Assays ›
Gene Symbol:
Rims2
LOC103692905
Gene Name:
regulating synaptic membrane exocytosis 2
regulating synaptic membrane exocytosis protein 2-like
Gene Aliases:
Nim2, Rim2
-
Chromosome Location:
Chr.7: 78092020 - 78594162 on Build Rnor_6.0
Chromosome Location:
Chr.7: 78671923 - 78677321 on Build Rnor_6.0
UniGene ID:
Rn.161948
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID
s138401
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rims2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053945.2
NP_446397.1
29
4534
NM_145881.2
NP_665888.1
6
856
XM_006241591.2
XP_006241653.1
27
4602
XM_006241592.2
XP_006241654.1
26
4536
XM_006241593.2
XP_006241655.1
28
4502
XM_006241594.2
XP_006241656.1
27
4460
XM_006241598.2
XP_006241660.1
27
7306
XM_008765437.1
XP_008763659.1
30
6804
XM_008765438.1
XP_008763660.1
28
4653
XM_008765439.1
XP_008763661.1
26
6498
XM_008765440.1
XP_008763662.1
24
4083
XM_008765441.1
XP_008763663.1
25
3990
XM_008765442.1
XP_008763664.1
24
3942
XM_008765443.1
XP_008763665.1
23
3875
XM_008765445.1
XP_008763667.1
7
1784
XM_008765446.1
XP_008763668.1
7
3717
XM_008765447.1
XP_008763669.1
27
4476
GenBank mRNA
AF199322.1
-
29
4534
AF199323.1
-
24
3925
AF199324.1
-
24
4066
AF199325.1
-
25
3973
AF199326.1
-
24
3925
AF199327.1
-
24
3925
AF199328.1
-
23
3745
AF199329.1
-
23
3859
AF199330.1
-
28
4486
AF199331.1
-
27
4444
AF199332.1
-
24
3925
AF199335.1
-
6
857
AF548738.1
-
27
3957
Gene Symbol: LOC103692905
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_008765484.1
XP_008763706.1
427
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
exocytosis
positive regulation of gene expression
calcium ion regulated exocytosis
regulation of exocytosis
cAMP-mediated signaling
insulin secretion
regulation of membrane potential
calcium ion-regulated exocytosis of neurotransmitter
positive regulation of inhibitory postsynaptic potential
positive regulation of excitatory postsynaptic potential
protein binding
Rab GTPase binding
protein domain specific binding
ion channel binding
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-4nm4f:80/100.66.76.134:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0