Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS15A SIRNA Assays ›
          Gene Symbol:
          Rps15a
          Gene Name:
          ribosomal protein S15a
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 187759855 - 187766734 on Build Rnor_6.0
          UniGene ID:
          Rn.2890
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s138505

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps15a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053982.1 NP_446434.1 1 120
          XM_006230097.1 XP_006230159.1 2 167
          XM_006230098.1 XP_006230160.1 2 210
          GenBank mRNA BC058452.1 - 2 145 6921126
          FQ210209.1 - 2 142
          FQ211215.1 - 2 145
          FQ212042.1 - 2 141
          FQ212257.1 - 2 139
          FQ214481.1 - 2 141
          FQ220927.1 - 2 137
          FQ220997.1 - 2 141
          FQ221041.1 - 2 141
          FQ221144.1 - 2 172
          FQ221174.1 - 2 139
          FQ221230.1 - 2 214
          FQ221308.1 - 2 143
          FQ221419.1 - 2 141
          FQ221518.1 - 2 141
          FQ221677.1 - 2 139
          FQ221720.1 - 2 210
          FQ221825.1 - 2 141
          FQ221840.1 - 2 141
          FQ221877.1 - 2 141
          FQ222252.1 - 2 152
          FQ222272.1 - 2 141
          FQ222274.1 - 2 141
          FQ222304.1 - 2 141
          FQ222376.1 - 2 141
          FQ222415.1 - 2 139
          FQ222418.1 - 2 143
          FQ222718.1 - 2 141
          FQ222721.1 - 2 139
          FQ222768.1 - 2 139
          FQ222892.1 - 2 142
          FQ222921.1 - 2 144
          FQ222922.1 - 2 210
          FQ223006.1 - 2 141
          FQ223095.1 - 2 141
          FQ223230.1 - 2 140
          FQ223341.1 - 2 141
          FQ223389.1 - 2 141
          FQ223433.1 - 2 141
          FQ223787.1 - 2 141
          FQ223930.1 - 263
          FQ224347.1 - 2 141
          FQ224383.1 - 2 141
          FQ228419.1 - 2 140
          FQ228429.1 - 2 141
          FQ228446.1 - 2 141
          FQ228475.1 - 2 142
          FQ228508.1 - 2 141
          FQ228609.1 - 2 141
          FQ228617.1 - 2 140
          FQ229004.1 - 2 142
          FQ229042.1 - 2 139
          FQ229063.1 - 2 140
          X77953.1 - 1 120

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          positive regulation of cell proliferation
          response to virus
          positive regulation of cell cycle
          structural constituent of ribosome
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0