Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pou2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228368.2 | XP_006228430.2 | 11 | 1056 | ||
XM_006228371.2 | XP_006228433.2 | 11 | 1074 | |||
XM_006228372.2 | XP_006228434.2 | 11 | 1056 | |||
XM_008758885.1 | XP_008757107.1 | 11 | 1122 | |||
GenBank mRNA | FQ225995.1 | - | 4 | 376 | ||
L25863.1 | - | 11 | 959 | |||
X67302.1 | - | 3 | 353 |