Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TCF3 SIRNA Assays ›
          Gene Symbol:
          TCF3
          Gene Name:
          transcription factor 3
          Gene Aliases:
          AGM8, E2A, E47, ITF1, TCF-3, VDIR, bHLHb21
          Chromosome Location:
          Chr.19: 1609290 - 1652590 on Build GRCh38
          UniGene ID:
          Hs.371282
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13874

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TCF3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136139.2 NP_001129611.1 13, 14 1211
          NM_003200.3 NP_003191.1 14, 15 1241
          XM_006722852.3 XP_006722915.1 14, 15 1416
          XM_006722853.3 XP_006722916.1 14, 15 1416
          XM_006722855.3 XP_006722918.1 14, 15 1414
          XM_006722856.3 XP_006722919.1 14, 15 1415
          XM_006722857.3 XP_006722920.1 14, 15 1415
          XM_006722858.3 XP_006722921.1 14, 15 1416
          XM_011528215.2 XP_011526517.1 13, 14 2714
          XM_011528216.2 XP_011526518.1 14, 15 1415
          XM_011528217.2 XP_011526519.1 14, 15 1418
          XM_011528220.2 XP_011526522.1 14, 15 1414
          XM_011528221.2 XP_011526523.1 14, 15 1415
          XM_011528225.2 XP_011526527.1 14, 15 1327
          XM_011528227.2 XP_011526529.1 12, 13 1173
          XM_017027177.1 XP_016882666.1 14, 15 1415
          XM_017027178.1 XP_016882667.1 14, 15 1415
          XM_017027180.1 XP_016882669.1 12, 13 1177
          XM_017027181.1 XP_016882670.1 14, 15 1327
          XM_017027182.1 XP_016882671.1 14, 15 1326
          XM_017027184.1 XP_016882673.1 14, 15 1419
          GenBank mRNA BC011665.2 - 11, 12 985
          BC110580.1 - 11, 12 1047 40007779
          BG284440.1 - 2, 3 138
          KF303591.1 - 11, 12 1019
          M31222.1 - 1241
          M31523.1 - 13, 14 1202
          X52078.1 - 10, 11 968

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          B cell lineage commitment
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          B cell differentiation
          immunoglobulin V(D)J recombination
          positive regulation of neuron differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of muscle cell differentiation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          protein binding
          transcription factor binding
          mitogen-activated protein kinase kinase kinase binding
          enhancer binding
          protein homodimerization activity
          bHLH transcription factor binding
          protein heterodimerization activity
          repressing transcription factor binding
          vitamin D response element binding
          E-box binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0