Search
Search

Rplp2
LOC100911575| Gene Symbol: Rplp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030021.1 | NP_001025192.1 | 3 | 180 | ||
| GenBank mRNA | BC099697.1 | - | 3 | 180 | 7934014 | |
| FQ217138.1 | - | 3 | 193 | |||
| FQ217658.1 | - | 3 | 194 | |||
| FQ221181.1 | - | 3 | 193 | |||
| FQ221478.1 | - | 3 | 196 | |||
| FQ221584.1 | - | 3 | 192 | |||
| FQ221984.1 | - | 3 | 194 | |||
| FQ222461.1 | - | 3 | 192 | |||
| FQ222498.1 | - | 3 | 193 | |||
| FQ223422.1 | - | 3 | 195 | |||
| FQ228678.1 | - | 3 | 194 | |||
| X15098.1 | - | 3 | 194 | |||
| Gene Symbol: LOC100911575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749013.2 | XP_003749061.1 | 3 | 415 | ||