Search
Search

TGFB3| Gene Symbol: TGFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003239.3 | NP_003230.1 | 7 | 2224 | ||
| GenBank mRNA | AK122902.1 | - | 4 | 1476 | ||
| AK313564.1 | - | 7 | 1305 | |||
| EF560714.1 | - | 7 | 1208 | |||
| J03241.1 | - | 7 | 1357 | |||
| X14149.1 | - | 7 | 1348 | |||