Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TGFB3 SIRNA Assays ›
          Gene Symbol:
          TGFB3
          Gene Name:
          transforming growth factor beta 3
          Gene Aliases:
          ARVD, ARVD1, LDS5, RNHF, TGF-beta3
          Chromosome Location:
          Chr.14: 75958097 - 75982022 on Build GRCh38
          UniGene ID:
          Hs.592317
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s14063

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TGFB3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003239.3 NP_003230.1 1 1416
          GenBank mRNA AK313564.1 - 1 497
          BC018503.1 - 1 387 4477175
          BT007287.1 - 1 287
          EF560714.1 - 1 400
          J03241.1 - 1 549
          X14149.1 - 1 540

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          response to hypoxia
          in utero embryonic development
          platelet degranulation
          transforming growth factor beta receptor signaling pathway
          SMAD protein import into nucleus
          salivary gland morphogenesis
          female pregnancy
          aging
          negative regulation of DNA replication
          negative regulation of cell proliferation
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          negative regulation of macrophage cytokine production
          cell growth
          positive regulation of bone mineralization
          negative regulation of transforming growth factor beta receptor signaling pathway
          mammary gland development
          response to progesterone
          positive regulation of collagen biosynthetic process
          response to laminar fluid shear stress
          wound healing
          regulation of cell proliferation
          odontogenesis
          uterine wall breakdown
          regulation of apoptotic process
          positive regulation of apoptotic process
          regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          response to estrogen
          ossification involved in bone remodeling
          cell-cell junction organization
          positive regulation of DNA replication
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          lung alveolus development
          digestive tract development
          embryonic neurocranium morphogenesis
          inner ear development
          positive regulation of protein secretion
          positive regulation of filopodium assembly
          positive regulation of stress fiber assembly
          positive regulation of cell division
          palate development
          face morphogenesis
          frontal suture morphogenesis
          positive regulation of SMAD protein import into nucleus
          SMAD protein signal transduction
          detection of hypoxia
          regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
          positive regulation of occluding junction disassembly
          type II transforming growth factor beta receptor binding
          cytokine activity
          protein binding
          growth factor activity
          type I transforming growth factor beta receptor binding
          type III transforming growth factor beta receptor binding
          identical protein binding
          protein heterodimerization activity
          transforming growth factor beta binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0