Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034917.2 | NP_001030089.2 | 16, 17 | 1507 | ||
GenBank mRNA | BC099806.1 | - | 16, 17 | 1505 | 7938236 |