Search
Search

Pcgf2| Gene Symbol: Pcgf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105836.2 | NP_001099306.1 | 9 | 1340 | ||
| XM_008767971.1 | XP_008766193.1 | 10 | 1299 | |||
| XM_008767972.1 | XP_008766194.1 | 10 | 1575 | |||
| XM_008767973.1 | XP_008766195.1 | 10 | 1645 | |||
| GenBank mRNA | BC167059.1 | - | 9 | 1340 | 7445511 | |