Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247470.2 | XP_006247532.1 | 7, 8 | 1872 | ||
XM_006247472.2 | XP_006247534.1 | 7, 8 | 1850 | |||
XM_008768223.1 | XP_008766445.1 | 7, 8 | 1931 | |||
XM_008768224.1 | XP_008766446.1 | 8, 9 | 1275 | |||
XM_008768225.1 | XP_008766447.1 | 7, 8 | 1909 |