Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dnajb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013209.1 | NP_001013227.1 | 3, 4 | 285 | ||
XM_006235863.2 | XP_006235925.1 | 3, 4 | 331 | |||
XM_006235864.2 | XP_006235926.1 | 4, 5 | 388 | |||
XM_006235866.2 | XP_006235928.1 | 3, 4 | 328 | |||
GenBank mRNA | BC078908.1 | - | 3, 4 | 285 | 7113862 | |
FQ214601.1 | - | 3, 4 | 286 |
Gene Symbol: LOC690183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001073579.4 | XP_001073579.1 | 1 | 279 |
Gene Symbol: RGD1565996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768672.1 | XP_008766894.1 | 1 | 196 |