Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TPMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000367.3 | NP_000358.1 | 4 | 458 | ||
XM_011514839.2 | XP_011513141.1 | 4 | 416 | |||
XM_017011241.1 | XP_016866730.1 | 5 | 752 | |||
GenBank mRNA | AF021876.1 | - | 3 | 281 | ||
AF021877.1 | - | 3 | 281 | |||
AK310420.1 | - | 4 | 352 | |||
BC005339.1 | - | 4 | 368 | 3829338 | ||
BC009596.1 | - | 4 | 367 | 4133029 | ||
BI598325.1 | - | 4 | 443 | |||
DB481604.1 | - | 4 | 462 | |||
S62904.1 | - | 3 | 346 | |||
U12387.1 | - | 4 | 357 |