Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: APCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001639.3 | NP_001630.1 | 2 | 403 | ||
GenBank mRNA | BC007039.1 | - | 2 | 403 | 3935086 | |
BC007058.1 | - | 2 | 403 | 3934872 | ||
BT006750.1 | - | 2 | 307 | |||
CR450313.1 | - | 2 | 307 | |||
FJ460512.1 | - | 2 | 403 | |||
X04608.1 | - | 2 | 403 |