Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rab3gap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040154.2 | NP_001035244.2 | 32 | 3779 | ||
XM_008769835.1 | 33 | 3764 | ||||
XM_008769836.1 | XP_008768058.1 | 31 | 5185 | |||
XM_008769837.1 | XP_008768059.1 | 30 | 3455 | |||
GenBank mRNA | BC086380.1 | - | 6 | 620 |