Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DNM1 SIRNA Assays ›
          Gene Symbol:
          DNM1
          Gene Name:
          dynamin 1
          Gene Aliases:
          DNM, EIEE31
          Chromosome Location:
          Chr.9: 128203355 - 128255248 on Build GRCh38
          UniGene ID:
          Hs.522413
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s146

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DNM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005336.2 NP_001005336.1 3 376
          NM_001288737.1 NP_001275666.1 3 376
          NM_001288738.1 NP_001275667.1 3 376
          NM_001288739.1 NP_001275668.1 3 376
          NM_004408.3 NP_004399.2 3 376
          XM_005251763.2 XP_005251820.1 3 376
          XM_005251768.2 XP_005251825.1 3 376
          XM_005251769.2 XP_005251826.1 3 376
          XM_006716992.2 XP_006717055.1 3 376
          XM_006716993.1 XP_006717056.1 3 376
          XM_011518335.2 XP_011516637.1 3 376
          XM_011518336.2 XP_011516638.1 3 376
          XM_017014369.1 XP_016869858.1 3 376
          XM_017014370.1 XP_016869859.1 3 376
          XM_017014371.1 XP_016869860.1 3 376
          XM_017014372.1 XP_016869861.1 3 376
          XM_017014373.1 XP_016869862.1 3 376
          GenBank mRNA AB209124.1 - 2 61
          AK225179.1 - 3 285
          AK295101.1 - 3 169
          AK296326.1 - 3 343
          AK316235.1 - 3 352
          BC050279.1 - 3 352 5284480
          BC063850.1 - 3 309 6184100
          DA221568.1 - 3 376
          DA510154.1 - 3 356
          L07807.1 - 3 326

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor internalization
          endocytosis
          receptor-mediated endocytosis
          endosome organization
          sensory perception of sound
          adult locomotory behavior
          ephrin receptor signaling pathway
          protein tetramerization
          synaptic transmission, GABAergic
          clathrin-mediated endocytosis
          toxin transport
          positive regulation of synaptic vesicle recycling
          GTPase activity
          protein binding
          GTP binding
          protein C-terminus binding
          protein kinase binding
          D2 dopamine receptor binding
          protein complex binding
          identical protein binding
          poly(A) RNA binding
          protein dimerization activity
          nitric-oxide synthase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0