Search
Search

Tsen34
LOC100909510| Gene Symbol: Tsen34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006968.1 | NP_001006969.1 | 710 | |||
| XM_006228017.2 | XP_006228079.1 | 1144 | ||||
| XM_006228018.2 | XP_006228080.1 | 718 | ||||
| XM_008758752.1 | XP_008756974.1 | 1323 | ||||
| GenBank mRNA | BC083767.1 | - | 710 | 7193201 | ||
| Gene Symbol: LOC100909510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748716.3 | XP_003748764.1 | 4 | 1080 | ||
| XM_003748717.3 | XP_003748765.1 | 4 | 718 | |||
| XM_006228075.2 | XP_006228137.1 | 4 | 1144 | |||
| XM_008758839.1 | XP_008757061.1 | 3 | 1323 | |||