Search
Search

Siglec5| Gene Symbol: Siglec5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106249.2 | NP_001099719.2 | 3 | 690 | ||
| XM_006228946.2 | XP_006229008.1 | 3 | 803 | |||
| XM_008759327.1 | XP_008757549.1 | 3 | 778 | |||
| XM_008759328.1 | XP_008757550.1 | 3 | 778 | |||
| XM_008759329.1 | XP_008757551.1 | 3 | 780 | |||