Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cdc42bpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130013.1 | NP_001123485.1 | 31 | 4239 | ||
XM_006230751.2 | XP_006230813.1 | 31 | 4178 | |||
XM_006230752.2 | XP_006230814.1 | 31 | 4105 | |||
XM_008760108.1 | XP_008758330.1 | 31 | 3889 |