Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Esrra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008511.2 | NP_001008511.2 | 6 | 877 | ||
XM_006230755.1 | XP_006230817.1 | 6 | 990 | |||
GenBank mRNA | AY280663.1 | - | 5 | 771 | ||
BC104701.1 | - | 6 | 877 |