Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011559.1 | NP_001011559.1 | 30 | 4186 | ||
XM_006256248.2 | XP_006256310.1 | 31 | 4376 | |||
XM_006256249.2 | XP_006256311.1 | 31 | 4373 | |||
GenBank mRNA | AB193179.1 | - | 30 | 4214 | ||
AY749166.1 | - | 30 | 4186 |