Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gpatch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024979.1 | NP_001020150.1 | 5 | 432 | ||
XM_008761129.1 | XP_008759351.1 | 6 | 963 | |||
XM_008761130.1 | XP_008759352.1 | 6 | 1013 | |||
XM_008761131.1 | XP_008759353.1 | 5 | 949 | |||
XM_008761132.1 | XP_008759354.1 | 6 | 1127 | |||
GenBank mRNA | BC093381.1 | - | 5 | 432 | 7388663 | |
FQ233538.1 | - | 2 | 154 |