Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APOE SIRNA Assays ›
          Gene Symbol:
          APOE
          Gene Name:
          apolipoprotein E
          Gene Aliases:
          AD2, APO-E, LDLCQ5, LPG
          Chromosome Location:
          Chr.19: 44905749 - 44909395 on Build GRCh38
          UniGene ID:
          Hs.654439
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1495

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APOE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000041.3 NP_000032.1 4 385
          NM_001302688.1 NP_001289617.1 4 467
          NM_001302689.1 NP_001289618.1 4 316
          NM_001302690.1 NP_001289619.1 4 416
          NM_001302691.1 NP_001289620.1 4 400
          GenBank mRNA AK314898.1 - 4 338
          BC003557.1 - 4 314 3355712
          BI550631.1 - 4 362
          BM042032.1 - 4 400
          BU848796.1 - 352
          CA771535.1 - 3 534
          HY056112.1 - 4 385
          HY074384.1 - 4 316
          HY074888.1 - 4 416
          K00396.1 - 4 329
          KU177910.1 - 3 269
          KU177911.1 - 3 269
          M12529.1 - 4 330
          X00199.1 - 2 80

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          retinoid metabolic process
          negative regulation of endothelial cell proliferation
          response to dietary excess
          triglyceride metabolic process
          cholesterol catabolic process
          cellular calcium ion homeostasis
          receptor-mediated endocytosis
          cytoskeleton organization
          G-protein coupled receptor signaling pathway
          nitric oxide mediated signal transduction
          synaptic transmission, cholinergic
          cholesterol metabolic process
          regulation of gene expression
          negative regulation of platelet activation
          positive regulation of cholesterol esterification
          positive regulation of cholesterol efflux
          long-chain fatty acid transport
          protein import
          virion assembly
          triglyceride catabolic process
          cGMP-mediated signaling
          negative regulation of blood coagulation
          regulation of axon extension
          positive regulation of cGMP biosynthetic process
          neuron projection regeneration
          regulation of Cdc42 protein signal transduction
          positive regulation of low-density lipoprotein particle receptor catabolic process
          cholesterol efflux
          phospholipid efflux
          very-low-density lipoprotein particle remodeling
          low-density lipoprotein particle remodeling
          high-density lipoprotein particle remodeling
          high-density lipoprotein particle assembly
          chylomicron remnant clearance
          high-density lipoprotein particle clearance
          very-low-density lipoprotein particle clearance
          lipoprotein metabolic process
          lipoprotein biosynthetic process
          lipoprotein catabolic process
          vasodilation
          cholesterol homeostasis
          negative regulation of MAP kinase activity
          negative regulation of neuron apoptotic process
          negative regulation of blood vessel endothelial cell migration
          reverse cholesterol transport
          positive regulation by host of viral process
          negative regulation of cholesterol biosynthetic process
          positive regulation of lipid biosynthetic process
          intracellular transport
          regulation of neuronal synaptic plasticity
          artery morphogenesis
          negative regulation of inflammatory response
          positive regulation of nitric-oxide synthase activity
          positive regulation of membrane protein ectodomain proteolysis
          maintenance of location in cell
          fatty acid homeostasis
          positive regulation of dendritic spine development
          negative regulation of canonical Wnt signaling pathway
          AMPA glutamate receptor clustering
          NMDA glutamate receptor clustering
          cellular oxidant detoxification
          regulation of beta-amyloid clearance
          negative regulation of neuron death
          positive regulation of postsynaptic membrane organization
          negative regulation of presynaptic membrane organization
          negative regulation of beta-amyloid formation
          positive regulation of dendritic spine maintenance
          positive regulation of phospholipid efflux
          positive regulation of lipid transport across blood brain barrier
          beta-amyloid binding
          lipid transporter activity
          protein binding
          phospholipid binding
          heparin binding
          lipid binding
          cholesterol binding
          antioxidant activity
          cholesterol transporter activity
          identical protein binding
          protein homodimerization activity
          metal chelating activity
          tau protein binding
          low-density lipoprotein particle receptor binding
          phosphatidylcholine-sterol O-acyltransferase activator activity
          very-low-density lipoprotein particle receptor binding
          lipoprotein particle binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0