Search
Search

ZNF7| Gene Symbol: ZNF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282795.1 | NP_001269724.1 | 5 | 469 | ||
| NM_001282797.1 | NP_001269726.1 | 4 | 309 | |||
| NM_003416.3 | NP_003407.1 | 5 | 436 | |||
| XM_006716654.2 | XP_006716717.1 | 3 | 200 | |||
| XM_006716656.3 | XP_006716719.1 | 3 | 189 | |||
| XM_011517291.2 | XP_011515593.1 | 5 | 483 | |||
| XM_011517292.2 | XP_011515594.1 | 5 | 817 | |||
| XM_011517293.2 | XP_011515595.1 | 5 | 816 | |||
| XM_011517294.2 | XP_011515596.1 | 5 | 595 | |||
| XM_011517295.2 | XP_011515597.1 | 5 | 376 | |||
| XM_011517296.2 | XP_011515598.1 | 5 | 387 | |||
| XM_011517297.2 | XP_011515599.1 | 3 | 236 | |||
| XM_017013813.1 | XP_016869302.1 | 5 | 480 | |||
| XM_017013814.1 | XP_016869303.1 | 4 | 782 | |||
| XM_017013815.1 | XP_016869304.1 | 4 | 779 | |||
| GenBank mRNA | AB209619.1 | - | 2 | 3825 | ||
| AK300001.1 | - | 5 | 404 | |||
| AK301561.1 | - | 4 | 270 | |||
| AK308002.1 | - | 5 | 428 | |||
| AK313745.1 | - | 5 | 376 | |||
| BC058923.1 | - | 5 | 380 | 4516166 | ||
| BJ989518.1 | - | 3 | 270 | |||
| BX647998.1 | - | 5 | 395 | |||
| DC301751.1 | - | 5 | 494 | |||
| M29580.1 | - | 5 | 537 | |||