Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC298111 SIRNA Assays ›
          Gene Symbol:
          LOC298111 LOC102548083 LOC103690101 LOC259244 LOC259246 LOC298116 LOC500473 Mup4 RGD1566134
          Gene Name:
          alpha2u globulin
          major urinary protein-like
          major urinary protein-like
          alpha-2u globulin PGCL3
          alpha-2u globulin PGCL1
          alpha-2u-globulin (L type)
          similar to alpha-2u globulin PGCL2
          major urinary protein 4
          similar to alpha-2u-globulin
          Gene Aliases:
          -
          -
          -
          -
          Mup
          -
          -
          -
          -
          Chromosome Location:
          Chr.5: 52651399 - 52655608 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 77245205 - 77299092 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 77723639 - 77725821 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 77404908 - 77408425 on Build Rnor_6.0
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.5: 77339109 - 77342362 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 77313449 - 77316803 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 77431489 - 77433910 on Build Rnor_6.0
          UniGene ID:
          Rn.195845
          -
          -
          Rn.86412
          Rn.224459
          Rn.86422
          -
          Rn.107064
          Rn.86422
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s151172

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC298111
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001024248.1 NP_001019419.1 321
          Gene Symbol: LOC102548083
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008763747.1 XP_008761969.1 388
          Gene Symbol: LOC103690101
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008763753.1 XP_008761975.1 4 321
          Gene Symbol: LOC259244
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_147212.1 NP_671745.1 360
          GenBank mRNA AB039824.1 - 360
          Gene Symbol: LOC259246
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_147214.2 NP_671747.1 4 367
          GenBank mRNA AB039822.1 - 4 360
          BC088109.1 - 4 362 7322607
          BC098654.1 - 4 367 7370970
          BC105816.1 - 4 364 7367233
          J00737.1 - 4 309
          M26835.1 - 4 339
          M26837.1 - 4 335
          M26838.1 - 4 332
          M27434.1 - 4 309
          U31287.1 - 4 374
          Gene Symbol: LOC298116
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001003409.1 NP_001003409.1 4 324
          GenBank mRNA M26836.1 - 4 324
          Gene Symbol: LOC500473
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006238221.2 XP_006238283.1 435
          Gene Symbol: Mup4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_198784.1 NP_942079.1 360
          GenBank mRNA AB039829.1 - 360
          Gene Symbol: RGD1566134
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_203512.2 NP_988843.2 360
          GenBank mRNA AB039823.1 - 360
          BC086943.1 - 372 7372252

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          energy reserve metabolic process
          insulin receptor signaling pathway
          aerobic respiration
          positive regulation of gene expression
          negative regulation of lipid storage
          positive regulation of glucose metabolic process
          heat generation
          glucose homeostasis
          locomotor rhythm
          negative regulation of gluconeogenesis
          positive regulation of lipid metabolic process
          negative regulation of transcription, DNA-templated
          negative regulation of lipid biosynthetic process
          positive regulation of protein kinase B signaling
          negative regulation of insulin secretion involved in cellular response to glucose stimulus
          mitochondrion morphogenesis
          cellular response to lipid
          transporter activity
          pheromone binding
          small molecule binding
          insulin-activated receptor activity
          odorant binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0