Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prepl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010951.1 | NP_001010951.1 | 13 | 1962 | ||
XM_006239697.2 | XP_006239759.1 | 14 | 2525 | |||
XM_006239698.2 | XP_006239760.2 | 14 | 2090 | |||
XM_006239699.2 | XP_006239761.2 | 14 | 2802 | |||
XM_008764456.1 | XP_008762678.1 | 14 | 2192 | |||
XM_008764457.1 | XP_008762679.1 | 14 | 2085 | |||
XM_008764458.1 | XP_008762680.1 | 13 | 1885 | |||
GenBank mRNA | BC089111.1 | - | 13 | 1962 | 7312962 | |
FQ213992.1 | - | 7 | 1001 |