Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106737.1 | NP_001100207.1 | 27 | 5841 | ||
XM_006240158.2 | XP_006240220.1 | 28 | 6001 | |||
XM_008764703.1 | XP_008762925.1 | 27 | 6134 | |||
XM_008764704.1 | XP_008762926.1 | 28 | 6001 | |||
XM_008764705.1 | XP_008762927.1 | 27 | 5911 | |||
XM_008764706.1 | XP_008762928.1 | 27 | 5911 | |||
XM_008764707.1 | XP_008762929.1 | 28 | 6001 | |||
XM_008764708.1 | XP_008762930.1 | 27 | 5911 | |||
XM_008764709.1 | XP_008762931.1 | 28 | 6001 | |||
XM_008764710.1 | XP_008762932.1 | 26 | 3788 |
Gene Symbol: LOC100911256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240147.2 | XP_006240209.1 | 5493 |