Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Acyp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106746.2 | NP_001100216.1 | 3 | 279 | ||
NM_001199167.1 | NP_001186096.1 | 3 | 211 | |||
NM_001199168.1 | NP_001186097.1 | 3 | 175 | |||
XM_006240308.2 | XP_006240370.1 | 3 | 330 | |||
GenBank mRNA | FM034312.1 | - | 3 | 175 | ||
FM062949.1 | - | 3 | 267 |