Search
Search

Prkcsh| Gene Symbol: Prkcsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106806.2 | NP_001100276.1 | 13 | 1223 | ||
| XM_006242633.2 | XP_006242695.1 | 13 | 1291 | |||
| XM_006242634.1 | XP_006242696.1 | 13 | 1356 | |||
| GenBank mRNA | BC161987.1 | - | 13 | 1223 | 9030870 | |