Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FZD5 SIRNA Assays ›
          Gene Symbol:
          FZD5
          Gene Name:
          frizzled class receptor 5
          Gene Aliases:
          C2orf31, HFZ5
          Chromosome Location:
          Chr.2: 207762586 - 207769419 on Build GRCh38
          UniGene ID:
          Hs.17631
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s15416
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FZD5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003468.3 NP_003459.2 2 1764
          GenBank mRNA AB043702.1 - 2 1739
          AK315338.1 - 1 1613
          BC114346.2 - 1 1407
          U43318.1 - 2 1674

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Matching Taqman Gene Expression Assay:


          Hs00361869_g1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic axis specification
          angiogenesis
          positive regulation of T cell cytokine production
          G-protein coupled receptor signaling pathway
          Wnt signaling pathway, calcium modulating pathway
          synapse assembly
          negative regulation of cell proliferation
          anterior/posterior axis specification, embryo
          neuron differentiation
          post-embryonic camera-type eye development
          positive regulation of interferon-gamma production
          T cell differentiation in thymus
          non-canonical Wnt signaling pathway
          positive regulation of JUN kinase activity
          Wnt signaling pathway involved in dorsal/ventral axis specification
          positive regulation of transcription from RNA polymerase II promoter
          cell maturation
          embryonic camera-type eye morphogenesis
          Spemann organizer formation
          canonical Wnt signaling pathway
          Wnt signaling pathway, planar cell polarity pathway
          apoptotic process involved in morphogenesis
          branching involved in labyrinthine layer morphogenesis
          syncytiotrophoblast cell differentiation involved in labyrinthine layer development
          labyrinthine layer blood vessel development
          chorionic trophoblast cell differentiation
          regulation of canonical Wnt signaling pathway
          cellular response to molecule of bacterial origin
          regulation of chorionic trophoblast cell proliferation
          regulation of mitophagy
          positive regulation of protein targeting to mitochondrion
          beta-catenin destruction complex disassembly
          regulation of bicellular tight junction assembly
          beta-amyloid binding
          G-protein coupled receptor activity
          protein binding
          Wnt-protein binding
          protein kinase binding
          ubiquitin protein ligase binding
          Wnt-activated receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0