Search
Search

Magea8
RGD1566060| Gene Symbol: Magea8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773233.1 | XP_008771455.1 | 4 | 4628 | ||
| XM_228890.3 | XP_228890.1 | 5 | 3096 | |||
| Gene Symbol: RGD1566060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773234.1 | XP_008771456.1 | 5 | 958 | ||
| XM_008773235.1 | XP_008771457.1 | 5 | 591 | |||
| XM_008773236.1 | XP_008771458.1 | 5 | 595 | |||